Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | flavin adenine dinucleotide synthetase 1 | FLAD1 |
FAD1, FADS, LSMFLAD, PP591 |
Sense | Chr. 1 - 154983294 - 154993111 on NCBI Build GRCh38 | |
| Downstream | lens epithelial protein | LENEP | LEP503 | 9.2kb | Sense | Chr. 1 - 154993586 - 154994315 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map