Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105373185 | LOC105373185 | -25.9kb | Anti-sense | Chr. X - 41275739 - 41276778 on NCBI Build GRCh38 | |
| Downstream | shisa family member 5 pseudogene | LOC643043 | 19.7kb | Sense | Chr. X - 41321413 - 41323574 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map