Assay Details
| Gene Symbol: LOC101928191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125851.1 | 1 - 2 | 41 | 61 | ||
| GenBank mRNA | BC033321.1 | - | 1 - 2 | 41 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | synaptonemal complex protein 2 like | SYCP2L |
C6orf177, NO145, dJ62D2.1 |
Anti-sense | Chr. 6 - 10886831 - 10974309 on NCBI Build GRCh38 | |
| Downstream | ELOVL fatty acid elongase 2 | ELOVL2 | SSC2 | 26.7kb | Sense | Chr. 6 - 10980759 - 11044391 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928191
Genomic Map