Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CKMT2 antisense RNA 1 | CKMT2-AS1 | Sense | Chr. 5 - 81237565 - 81301569 on NCBI Build GRCh38 | ||
| Downstream | zinc finger CCHC-type containing 9 | ZCCHC9 | PPP1R41 | 2.9kb | Anti-sense | Chr. 5 - 81301583 - 81313147 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map