Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | potassium two pore domain channel subfamily K member 15 | KCNK15 |
K2p15.1, KCNK11, KCNK14, KT3.3, TASK-5, TASK5, dJ781B1.1 |
Sense | Chr. 20 - 44745605 - 44752313 on NCBI Build GRCh38 | |
| Downstream | regulating synaptic membrane exocytosis 4 | RIMS4 |
C20orf190, RIM 4, RIM-4, RIM4, RIM4-gamma, RIM4gamma |
4.6kb | Anti-sense | Chr. 20 - 44751804 - 44810338 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map