Assay Details
| Gene Symbol: LOC105371708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC034720.1 | - | 3 - 4 | 410 | 106 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BE777946.1 | - | 390 | Chr. - - [-] on Build | 106 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LYR motif containing 9 | LYRM9 |
C17orf108, HSD24 |
-36.2kb | Sense | Chr. 17 - 27878314 - 27893383 on NCBI Build GRCh38 |
| Downstream | nemo like kinase | NLK | 111.9kb | Anti-sense | Chr. 17 - 28042647 - 28205140 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105371708
Genomic Map