Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1909 | MIR1909 |
MIRN1909, hsa-mir-1909, mir-1909 |
-5.9kb | Anti-sense | Chr. 19 - 1816159 - 1816238 on NCBI Build GRCh38 |
| Downstream | Kruppel like factor 16 | KLF16 |
BTEB4, DRRF, NSLP2 |
27.9kb | Anti-sense | Chr. 19 - 1852399 - 1863565 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map