Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1280 | LINC01280 | Anti-sense | Chr. 2 - 216589993 - 216606938 on NCBI Build GRCh38 | ||
| Downstream | insulin like growth factor binding protein 2 | IGFBP2 |
IBP2, IGF-BP53 |
22.2kb | Sense | Chr. 2 - 216632828 - 216664436 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map