Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | single-minded family bHLH transcription factor 2 | SIM2 |
HMC13F06, HMC29C01, SIM, bHLHe15 |
-133.0kb | Anti-sense | Chr. 21 - 36699117 - 36750219 on NCBI Build GRCh38 |
| Downstream | ripply transcriptional repressor 3 | RIPPLY3 | DSCR6 | 121.7kb | Anti-sense | Chr. 21 - 37006150 - 37019659 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map