Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADP ribosylation factor like GTPase 14 effector protein like | ARL14EPL | -174.1kb | Sense | Chr. 5 - 116051140 - 116060023 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101927190 | LOC101927190 | 130.9kb | Anti-sense | Chr. 5 - 116367416 - 116374570 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map