Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1395 | LINC01395 | -224.3kb | Sense | Chr. 11 - 129612116 - 129617269 on NCBI Build GRCh38 | |
| Downstream | nuclear factor related to kappaB binding protein | NFRKB | INO80G | 19.8kb | Sense | Chr. 11 - 129863773 - 129895596 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map