Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | antagonist of mitotic exit network 1 homolog | AMN1 | Anti-sense | Chr. 12 - 31671137 - 31729178 on NCBI Build GRCh38 | ||
| Downstream |
H3 histone, family 3C |
H3F3C | H3.5 | 103.5kb | Anti-sense | Chr. 12 - 31791185 - 31792241 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map