Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3921 | MIR3921 | -50.5kb | Sense | Chr. 3 - 99964314 - 99964398 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105374010 | LOC105374010 | 6.1kb | Anti-sense | Chr. 3 - 100021012 - 100034855 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map