Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADAM metallopeptidase domain 30 | ADAM30 | svph4 | -12.7kb | Sense | Chr. 1 - 119893533 - 119896524 on NCBI Build GRCh38 |
| Downstream | notch 2 | NOTCH2 |
AGS2, HJCYS, hN2 |
940.0 | Sense | Chr. 1 - 119911553 - 120073449 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map