Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | translation regulatory long non-coding RNA 1 | TRERNA1 |
LINC00651, treRNA |
-43.2kb | Sense | Chr. 20 - 50040707 - 50041629 on NCBI Build GRCh38 |
| Downstream | transmembrane protein 189 | TMEM189 | KUA | 37.8kb | Sense | Chr. 20 - 50123737 - 50153798 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map