Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PIK3CD antisense RNA 2 | PIK3CD-AS2 | Sense | Chr. 1 - 9672426 - 9687574 on NCBI Build GRCh38 | ||
| Downstream | calsyntenin 1 | CLSTN1 |
ALC-ALPHA, CDHR12, CST-1, CSTN1, PIK3CD, XB31alpha, alcalpha1, alcalpha2 |
41.5kb | Sense | Chr. 1 - 9729021 - 9824526 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map