Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DiGeorge syndrome critical region gene 11 (non-protein coding) | DGCR11 | DGS-D | -39.3kb | Sense | Chr. 22 - 19046162 - 19048375 on NCBI Build GRCh38 |
| Downstream | DiGeorge syndrome critical region gene 14 | DGCR14 |
DGCR13, DGS-H, DGS-I, DGSH, DGSI, ES2, Es2el |
40.9kb | Sense | Chr. 22 - 19130279 - 19144726 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map