Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
Smith-Magenis syndrome chromosome region, candidate 2 (non-protein coding) |
SMCR2 | TCONS_00025215 | -527.0 | Anti-sense | Chr. 17 - 17674026 - 17677688 on NCBI Build GRCh38 |
| Downstream | RAI1 antisense RNA 1 | RAI1-AS1 | 74.1kb | Anti-sense | Chr. 17 - 17759154 - 17766302 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map