Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Yip1 domain family member 4 | YIPF4 |
FinGER4, Nbla11189 |
-49.4kb | Anti-sense | Chr. 2 - 32277889 - 32307929 on NCBI Build GRCh38 |
| Downstream | microRNA 558 | MIR558 |
MIRN558, hsa-mir-558 |
174.1kb | Anti-sense | Chr. 2 - 32532153 - 32532246 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map