Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ring finger protein 4 pseudogene | LOC100419583 | Sense | Chr. 15 - 44728988 - 44732851 on NCBI Build GRCh38 | ||
| Downstream | tripartite motif containing 69 | TRIM69 |
HSD-34, HSD34, RNF36, Trif |
2.4kb | Sense | Chr. 15 - 44736362 - 44767829 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map