Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | interleukin 17F | IL17F |
CANDF6, IL-17F, ML-1, ML1 |
-38.6kb | Anti-sense | Chr. 6 - 52236681 - 52245689 on NCBI Build GRCh38 |
| Downstream | progestin and adipoQ receptor family member 8 | PAQR8 |
C6orf33, LMPB1, MPRB |
76.5kb | Sense | Chr. 6 - 52362128 - 52407777 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map