Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein L17 pseudogene 10 | RPL17P10 | RPL17_2_309 | -37.8kb | Anti-sense | Chr. 2 - 201102493 - 201102990 on NCBI Build GRCh38 |
| Downstream | caspase 10 | CASP10 |
ALPS2, FLICE2, MCH4 |
41.3kb | Sense | Chr. 2 - 201182881 - 201229406 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map