Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microtubule affinity regulating kinase 2 pseudogene 9 | MARK2P9 | -140.2kb | Anti-sense | Chr. 10 - 92418661 - 92419606 on NCBI Build GRCh38 | |
| Downstream | kinesin family member 11 | KIF11 |
EG5, HKSP, KNSL1, MCLMR, TRIP5 |
31.9kb | Anti-sense | Chr. 10 - 92593068 - 92655395 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map