Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glutathione S-transferase mu 5 | GSTM5 |
GSTM5-5, GTM5 |
-18.7kb | Anti-sense | Chr. 1 - 109711751 - 109718268 on NCBI Build GRCh38 |
| Downstream | EPS8 like 3 | EPS8L3 | EPS8R3 | 12.6kb | Sense | Chr. 1 - 109750080 - 109764022 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map