Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 11 open reading frame 84 | C11orf84 | -39.0kb | Sense | Chr. 11 - 63813374 - 63827718 on NCBI Build GRCh38 | |
| Downstream | REST corepressor 2 | RCOR2 | 43.4kb | Anti-sense | Chr. 11 - 63911221 - 63916844 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map