Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SPATA31 subfamily D member 1 | SPATA31D1 | FAM75D1 | -548.5kb | Anti-sense | Chr. 9 - 81987404 - 81995256 on NCBI Build GRCh38 |
| Downstream | RAS and EF-hand domain containing | RASEF | RAB45 | 435.4kb | Sense | Chr. 9 - 82979585 - 83063177 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map