Assay Details
| Gene Symbol: RCSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322923.1 | NP_001309852.1 | 6 | 3925 | 87 | |
| NM_001322924.1 | NP_001309853.1 | 5 | 3853 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cellular repressor of E1A stimulated genes 1 | CREG1 | CREG | -151.9kb | Anti-sense | Chr. 1 - 167541013 - 167553819 on NCBI Build GRCh38 |
| Downstream | myelin protein zero like 1 | MPZL1 |
MPZL1b, PZR, PZR1b, PZRa, PZRb |
14.7kb | Sense | Chr. 1 - 167721950 - 167791919 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RCSD1
Genomic Map