Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4316 | MIR4316 | -71.5kb | Sense | Chr. 17 - 77396984 - 77397054 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC400622 | LOC400622 | 55.8kb | Anti-sense | Chr. 17 - 77526571 - 77536598 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map