| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001271785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001271786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001271787.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001271788.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001271789.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001271790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001271785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001271786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001271787.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001271788.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001271789.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001271790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
NM_001378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200169861.hg.1 |
TC0200009959.hg.1 |
Clariom D TC |