Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 27 | LRRC27 | Sense | Chr. 10 - 132330063 - 132381506 on NCBI Build GRCh38 | ||
| Downstream | PWWP domain containing 2B | PWWP2B |
PWWP2, bA432J24.1, pp8607 |
51.0kb | Sense | Chr. 10 - 132397196 - 132417863 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map