Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ZFP69 zinc finger protein B | ZFP69B |
ZKSCAN23B, ZNF643, ZSCAN54B |
-9.3kb | Sense | Chr. 1 - 40450068 - 40466765 on NCBI Build GRCh38 |
| Downstream | ZFP69 zinc finger protein | ZFP69 |
ZFP69A, ZKSCAN23A, ZNF642, ZSCAN54A |
2.9kb | Sense | Chr. 1 - 40477220 - 40496343 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map