Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929577 | LOC101929577 | -19.2kb | Sense | Chr. 4 - 106003317 - 106022478 on NCBI Build GRCh38 | |
| Downstream | TBC1 domain containing kinase | TBCK |
HSPC302, IHPRF3, TBCKL |
938.0 | Sense | Chr. 4 - 106044317 - 106316704 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map