| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007294.hg.1 |
ENST00000565748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007294.hg.1 |
ENST00000565748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007294.hg.1 |
ENST00000565748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007294.hg.1 |
ENST00000569313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007294.hg.1 |
ENST00000569313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007294.hg.1 |
ENST00000565748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007294.hg.1 |
ENST00000565748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007294.hg.1 |
ENST00000565748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007294.hg.1 |
ENST00000569313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007294.hg.1 |
ENST00000569313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500174243.hg.1 |
NM_001098272.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174243.hg.1 |
NM_002130.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174243.hg.1 |
NM_001098272.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174243.hg.1 |
NM_002130.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174243.hg.1 |
TC0500010635.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500174241.hg.1 |
NM_001098272.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174241.hg.1 |
NM_002130.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174241.hg.1 |
NM_001098272.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174241.hg.1 |
NM_002130.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174241.hg.1 |
TC0500010635.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500174242.hg.1 |
NM_001098272.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174242.hg.1 |
NM_002130.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174242.hg.1 |
NM_001098272.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174242.hg.1 |
NM_002130.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174242.hg.1 |
TC0500010635.hg.1 |
Clariom D TC |