Assay Details
| Gene Symbol: ZKSCAN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278119.1 | NP_001265048.1 | 7 | 6767 | 95 | |
| NM_001278121.1 | NP_001265050.1 | 7 | 6714 | 95 | ||
| NM_001278122.1 | NP_001265051.1 | 6 | 6687 | 95 | ||
| NM_006298.3 | NP_006289.2 | 6 | 6673 | 95 | ||
| XM_011514870.2 | XP_011513172.1 | 6 | 6762 | 95 | ||
| XM_011514873.2 | XP_011513175.1 | 7 | 6873 | 95 | ||
| XM_011514874.2 | XP_011513176.1 | 5 | 6170 | 95 | ||
| XM_017011266.1 | XP_016866755.1 | 6 | 6677 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ZSCAN16 antisense RNA 1 | ZSCAN16-AS1 | -20.5kb | Anti-sense | Chr. 6 - 28121795 - 28137316 on NCBI Build GRCh38 | |
| Downstream | zinc finger protein 192 pseudogene 1 | ZNF192P1 |
ZNF389, ZNF389P, dJ265C24.4 |
2.3kb | Sense | Chr. 6 - 28161761 - 28169595 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

ZKSCAN8
Genomic Map