Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HEAT repeat containing 4 | HEATR4 | -9.3kb | Sense | Chr. 14 - 73478484 - 73634418 on NCBI Build GRCh38 | |
| Downstream | acyl-CoA thioesterase 4 | ACOT4 |
PTE-Ib, PTE1B, PTE2B |
21.9kb | Anti-sense | Chr. 14 - 73591528 - 73595766 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map