Assay Details
| Gene Symbol: GLB1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011542567.2 | XP_011540869.1 | 18 - 19 | 2383 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | acyl-CoA dehydrogenase family member 8 | ACAD8 |
ACAD-8, ARC42 |
-52.8kb | Sense | Chr. 11 - 134253532 - 134265858 on NCBI Build GRCh38 |
| Downstream | galactosidase beta 1 like 2 | GLB1L2 |
MST114, MSTP114 |
10.6kb | Sense | Chr. 11 - 134331874 - 134376324 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

GLB1L3
Genomic Map