| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000608735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000608735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000608735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000608735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000608735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000609836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009284.hg.1 |
ENST00000609836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000422048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000436019.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000608735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000608735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000608735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000608735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000608735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000609836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009284.hg.1 |
ENST00000609836.1 |
Ensembl |