Assay Details
| Gene Symbol: NCAPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281710.1 | NP_001268639.1 | 18 | 5547 | 85 | |
| NM_001281711.1 | NP_001268640.1 | 18 | 5508 | 85 | ||
| NM_001281712.1 | NP_001268641.1 | 17 | 5327 | 85 | ||
| NM_015341.4 | NP_056156.2 | 18 | 5580 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | inositol 1,4,5-trisphosphate receptor interacting protein-like 1 | ITPRIPL1 | KIAA1754L | -46.3kb | Sense | Chr. 2 - 96325324 - 96330507 on NCBI Build GRCh38 |
| Downstream | neuralized E3 ubiquitin protein ligase 3 | NEURL3 | LINCR | 120.6kb | Anti-sense | Chr. 2 - 96497643 - 96508157 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

NCAPH
Genomic Map