Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929371 | LOC101929371 | -16.7kb | Anti-sense | Chr. 20 - 9986333 - 10007116 on NCBI Build GRCh38 | |
| Downstream | ankyrin repeat and EF-hand domain containing 1 | ANKEF1 | ANKRD5 | 8.3kb | Anti-sense | Chr. 20 - 10034999 - 10056764 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map