Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC349160 | LOC349160 | -62.4kb | Anti-sense | Chr. 7 - 136898773 - 137164341 on NCBI Build GRCh38 | |
| Downstream | pleiotrophin | PTN |
HARP, HB-GAM, HBBM, HBGF-8, HBGF8, HBNF, HBNF-1, NEGF1, OSF-1 |
164.0 | Anti-sense | Chr. 7 - 137227341 - 137343865 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map