Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928271 | LOC101928271 | -180.3kb | Sense | Chr. 2 - 20859771 - 20861661 on NCBI Build GRCh38 | |
| Downstream | tudor domain containing 15 | TDRD15 | 64.3kb | Sense | Chr. 2 - 21123932 - 21147325 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map