Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | voltage dependent anion channel 1 | VDAC1 |
PORIN, VDAC-1 |
-53.1kb | Sense | Chr. 5 - 133971875 - 134070987 on NCBI Build GRCh38 |
| Downstream | uncharacterized protein C7orf73-like | LOC101060024 | 20.1kb | Sense | Chr. 5 - 134081411 - 134082833 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map