Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | junction adhesion molecule like | JAML |
AMICA, AMICA1, CREA7-1, CREA7-4, Gm638 |
Sense | Chr. 11 - 118193727 - 118225094 on NCBI Build GRCh38 | |
| Downstream | myelin protein zero like 3 | MPZL3 | 24.9kb | Sense | Chr. 11 - 118226690 - 118252368 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map