Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cyclin dependent kinase like 3 | CDKL3 | NKIAMRE | -12.3kb | Anti-sense | Chr. 5 - 134298424 - 134371159 on NCBI Build GRCh38 |
| Downstream | CDKN2A interacting protein N-terminal like | CDKN2AIPNL | 16.1kb | Anti-sense | Chr. 5 - 134402065 - 134414625 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map