Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 38 member 9 | SLC38A9 | URLC11 | Sense | Chr. 5 - 55625845 - 55712344 on NCBI Build GRCh38 | |
| Downstream | DEAD-box helicase 4 | DDX4 | VASA | 102.9kb | Anti-sense | Chr. 5 - 55737994 - 55817146 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map