Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis specific serine kinase 3 | TSSK3 |
SPOGA3, STK22C, STK22D, TSK3 |
-7.0kb | Anti-sense | Chr. 1 - 32362261 - 32364325 on NCBI Build GRCh38 |
| Downstream | zinc finger and BTB domain containing 8B | ZBTB8B | ZNF916B | 91.7kb | Anti-sense | Chr. 1 - 32465057 - 32487859 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map