Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | eukaryotic translation initiation factor 3 subunit F | EIF3F |
EIF3S5, eIF3-p47 |
-1.2kb | Sense | Chr. 11 - 7987320 - 7996171 on NCBI Build GRCh38 |
| Downstream | cancer susceptibility candidate 23 (non-protein coding) | CASC23 | LINC01464 | 11.8kb | Anti-sense | Chr. 11 - 8011278 - 8016509 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map