Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | olfactomedin 1 | OLFM1 |
AMY, NOE1, NOELIN1, OlfA |
Sense | Chr. 9 - 135075243 - 135121184 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC401557 | LOC401557 | 86.1kb | Sense | Chr. 9 - 135181609 - 135187306 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map