Assay Details
| Gene Symbol: SFT2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090795.1 | - | 1 | 353 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | proline rich 18 | PRR18 | -11.3kb | Sense | Chr. 6 - 166305680 - 166308383 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100289495 | LOC100289495 | 20.9kb | Anti-sense | Chr. 6 - 166342631 - 166351469 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SFT2D1
Genomic Map