Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myosin light chain 12B | MYL12B |
MLC-B, MRLC2 |
-48.4kb | Anti-sense | Chr. 18 - 3262113 - 3278284 on NCBI Build GRCh38 |
| Downstream | TGFB induced factor homeobox 1 | TGIF1 |
HPE4, TGIF |
81.2kb | Anti-sense | Chr. 18 - 3411927 - 3458412 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map